missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Quizalofop, TRC
Specifications
Specifications
| Kemiskt namn eller material | Quizalofop free acid |
| CAS | 76578-12-6 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C17 H13 Cl N2 O4 |
| Kvantitet | 100 mg |
| Synonym | Quizalofop (Free Acid), Quizalofop, Propanoic acid','2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]- (9CI','ACI), 2-[4-[(6-Chloro-2-quinoxalinyl)oxy]phenoxy]propanoic acid (ACI), Pilot, Quizalofop, Xylafop |
| InChI formel | InChI=1S/C17H13ClN2O4/c1-10(17(21)22)23-12-3-5-13(6-4-12)24-16-9-19-15-8-11(18)2-7-14(15)20-16/h2-10H,1H3,(H,21,22) |
| LEDER | CC(Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)O |
| IUPAC-namn | 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoic acid |
| Molekylvikt (g/mol) | 344.75 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?