Läs mer
(R)-1,2-Propylene Carbonate, TRC
Produktkod. 30424752
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30424752 5 g
1 options
Produktkod. 30424752 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8367505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-1,2-Propylene Carbonate
CAS 16606-55-6
Molekylformel C4 H6 O3
Kvantitet 5 g
Synonym 1,3-Dioxolan-2-one','4-methyl-','(4R)-, 1,3-Dioxolan-2-one','4-methyl-','(R)-, Carbonic acid','cyclic propylene ester','(+)- (8CI), (4R)-4-Methyl-1,3-dioxolan-2-one, (+)-(4R)-4-methyl-1,3-dioxolan-2-one, (+)-1-Methyl-2,4-dioxolan-3-one, (R)-(+)-1,2-Propylene carbonate, (R)-1,2-Propylene carbonate, (R)-Propylene carbonate
InChI formel InChI=1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3/t3-/m1/s1
LEDER C[C@@H]1COC(=O)O1
IUPAC-namn (4R)-4-methyl-1,3-dioxolan-2-one
Molekylvikt (g/mol) 102.09
Formel vikt 102.0317