missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Benzyl-3-pyrrolidinol, 98%, Thermo Scientific™

Catalog Number p-4380049
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
101930-07-8
C11H15NO
177.25
YQMXOIAIYXXXEE-LLVKDONJSA-N
r-+-1-benzyl-3-pyrrolidinol, r-3-hydroxy-1-benzylpyrrolidine, 3r-1-benzylpyrrolidin-3-ol, r-1-benzyl-3-pyrrolidinol, r-1-benzylpyrrolidin-3-ol, r-1-n-benzyl-3-hydroxypyrrolidine, r-1-benzyl-3-hydroxypyrrolidine, r-n-benzyl-3-hydroxypyrrolidine, r-1-benzyl-pyrrolidin-3-ol, r-bhp
643472
(3R)-1-benzylpyrrolidin-3-ol
C1CN(CC1O)CC2=CC=CC=C2
This item is not returnable. View return policy
101930-07-8
C11H15NO
177.25
YQMXOIAIYXXXEE-LLVKDONJSA-N
r-+-1-benzyl-3-pyrrolidinol, r-3-hydroxy-1-benzylpyrrolidine, 3r-1-benzylpyrrolidin-3-ol, r-1-benzyl-3-pyrrolidinol, r-1-benzylpyrrolidin-3-ol, r-1-n-benzyl-3-hydroxypyrrolidine, r-1-benzyl-3-hydroxypyrrolidine, r-n-benzyl-3-hydroxypyrrolidine, r-1-benzyl-pyrrolidin-3-ol, r-bhp
643472
(3R)-1-benzylpyrrolidin-3-ol
C1CN(CC1O)CC2=CC=CC=C2
Boiling Point 88°C to 90°C (1.0 Torr)
Assay Percent Range 97.5% min. (GC)
Quantity 1 g
Specific Rotation + 4
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material (R)-(+)-1-Benzyl-3-pyrrolidinol
compliance-icons
Product Identifier
  • (R)-(+)-1-Benzyl-3-pyrrolidinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'