Learn More
(R)-1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
(R)-1-Boc-3-hydroxypiperidine has been used as a reactant for the synthesis of piperidinyl and pyrrolidinyl butyrates. It has also been used for the synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-1-Boc-3-hydroxypiperidine has been used as a reactant for the synthesis of piperidinyl and pyrrolidinyl butyrates. It has also been used for the synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
(R)-1-Boc-3-hydroxypiperidine has been used as a reactant for the synthesis of piperidinyl and pyrrolidinyl butyrates. It has also been used for the synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Melting Point | 46°C to 50°C |
Flash Point | 104°C (219°F) |
Quantity | 1 g |
Optical Rotation | −23° (c=5 in Methanol) |
Formula Weight | 201.27 |
Percent Purity | 97% |
Chemical Name or Material | (R)-1-Boc-3-hydroxypiperidine |