Läs mer
(R)-1-Hydroxy-1-phenylpropanone, TRC
Produktkod. 17994225
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994225 100 mg
17970695 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17994225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH949610100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1798-60-3
Molekylformel C9 H10 O2
Molekylvikt (g/mol) 150.17
Synonym 2-Propanone, 1-hydroxy-1-phenyl-, (R)-, (1R)-1-Hydroxy-1-phenyl-2-propanone, (-)-(1R)-1-Hydroxy-1-phenyl-2-propanone, (1R)-1-Phenyl-1-hydroxypropan-2-one, (R)-1-Hydroxy-1-phenyl-2-propanone, (R)-1-Hydroxy-1-phenylpropanone, (R)-Hydroxy-1-Phenyl-2-propanedione, (R)-Phenylacetylcarbinol, D-(-)-1-Hydroxy-1-phenylpropanone, D-(-)-1-Phenyl-1-hydroxy-2-propanone, D-(-)-Phenylacetylcarbinol, PAC, l-Phenylacetylcarbinol, Ephedrine Hydrochloride Imp. A (EP)
IUPAC-namn (1R)-1-hydroxy-1-phenylpropan-2-one
LEDER CC(=O)[C@H](O)c1ccccc1
Specifikationer
Kemiskt namn eller material (R)-1-Hydroxy-1-phenylpropanone
Kvantitet 100 mg
InChI formel InChI=1S/C9H10O2/c1-7(10)9(11)8-5-3-2-4-6-8/h2-6,9,11H,1H3/t9-/m0/s1
Formel vikt 150.0681