Learn More
(R)-(+)-1-Phenylethyl isocyanate, 99%, Thermo Scientific Chemicals
Catalog Number p-7045335
Click to view available options
Quantity:
1 g
5 g
33375-06-3
C9H9NO
147.177
MFCD00063015
JJSCUXAFAJEQGB-MRVPVSSYSA-N
r-+-1-phenylethyl isocyanate, 1r-1-isocyanatoethyl benzene, r-+-alpha-methylbenzyl isocyanate, benzene, 1r-1-isocyanatoethyl, isocyanic acid r-+-1-phenylethyl ester, isocyanic acid r-+-alpha-methylbenzyl ester, isocyanic acid-r-+-methylbenzyl ester, 1-isocyanatoethyl benzene #, +-alpha-methylbenzyl isocyanate, pubchem8096
7018262
[(1R)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
33375-06-3
C9H9NO
147.177
MFCD00063015
JJSCUXAFAJEQGB-MRVPVSSYSA-N
r-+-1-phenylethyl isocyanate, 1r-1-isocyanatoethyl benzene, r-+-alpha-methylbenzyl isocyanate, benzene, 1r-1-isocyanatoethyl, isocyanic acid r-+-1-phenylethyl ester, isocyanic acid r-+-alpha-methylbenzyl ester, isocyanic acid-r-+-methylbenzyl ester, 1-isocyanatoethyl benzene #, +-alpha-methylbenzyl isocyanate, pubchem8096
7018262
[(1R)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.
Solubility
Decomposes in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.
TRUSTED_SUSTAINABILITY
Density 1.045
Boiling Point 55°C to 56°C (2.5 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.5124
Quantity 1 g
UN Number UN2206
Beilstein 3196930
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Optical Rotation +10.5° (Neat)
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material (R)-(+)-1-Phenylethyl isocyanate