Läs mer
Tocris Bioscience™ R 121919 Hydrochloride
Beskrivning
R 121919 hydrochloride is a high affinity corticotropin releasing factor receptor1 (CRF1) antagonist (Ki = 3.5 nM). Reduces stress-induced elevations of plasma ACTH. Anxiolytic and antidepressant in vivo. Orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-[6-(Dimethylamino)-4-methyl-3-pyridinyl]-2,5-dimethyl-N,N-dipropylpyrazolo[1,5-a]pyrimidin-7-amine hydrochloride |
| CAS | 195055-66-4 |
| Kvantitet | 10 mg |
| Molekylformel | C22H32N6.HCl |
| Formel vikt | 416.99 |
| Renhet | 98% |