Läs mer
(R)-2-Amino-3-(4-methoxyphenyl)propanoic Acid, TRC
Produktkod. 30334780
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30334780 1 g
1 options
Produktkod. 30334780 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA8982031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R)-2-Amino-3-(4-methoxyphenyl)propanoic Acid
CAS 39878-65-4
Molekylformel C10 H13 N O3
Kvantitet 1 g
Synonym D-Tyrosine, O-methyl-, O-Methyl-D-tyrosine, (2R)-2-Amino-3-(4-methoxyphenyl)propanoic acid, (R)-2-Amino-3-(4-methoxyphenyl)propionic acid, D-4-Methoxyphenylalanine, D-O-Methyltyrosine
InChI formel InChI=1S/C10H13NO3/c1-14-8-4-2-7(3-5-8)6-9(11)10(12)13/h2-5,9H,6,11H2,1H3,(H,12,13)/t9-/m1/s1
LEDER COc1ccc(C[C@@H](N)C(=O)O)cc1
IUPAC-namn (2R)-2-amino-3-(4-methoxyphenyl)propanoic acid
Molekylvikt (g/mol) 195.215
Formel vikt 195.09