Läs mer
(R)-2-Bromobutyric Acid, TRC
Produktkod. 30179787
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179787 500 mg
30179854 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179787 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB545000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2681-94-9
Molekylformel C4 H7 Br O2
Molekylvikt (g/mol) 167
Synonym Butanoic acid, 2-bromo-, (2R)- (9CI, ACI), (2R)-2-Bromobutanoic acid (ACI), Butanoic acid, 2-bromo-, (R)- (ZCI), Butyric acid, 2-bromo-, D- (8CI), (2R)-2-Bromobutanoic acid, (R)-2-Bromobutanoic acid, (R)-2-Bromobutyric acid, (R)-2-Bromobutyric acid, (R)-α-Bromobutyric acid, D-2-Bromobutanoic acid, D-2-Bromobutyric acid
IUPAC-namn (2R)-2-bromobutanoic acid
LEDER CC[C@@H](Br)C(=O)O
Specifikationer
Kemiskt namn eller material (R)-2-Bromobutyric Acid
Kvantitet 500 mg
InChI formel InChI=1S/C4H7BrO2/c1-2-3(5)4(6)7/h3H,2H2,1H3,(H,6,7)/t3-/m1/s1
Formel vikt 165.9629