Läs mer
(R)-(-)-2-Oxothiazolidine-4-carboxylic Acid, TRC
Produktkod. 30424317
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30424317 5 g
1 options
Produktkod. 30424317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8705055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-(-)-2-Oxothiazolidine-4-carboxylic Acid
CAS 19771-63-2
Molekylformel C4H5NO3S
Kvantitet 5 g
Synonym (4R)-2-Oxo-4-thiazolidinecarboxylic Acid, (-)-2-Oxo-4-thiazolidinecarboxylic Acid, 2-Oxo-4-thiazolidinecarboxylic Acid, L-2-Oxo-4-thiazolidinecarboxylic Acid, 4R)-2-Oxo-1,3-thiazolidine-4-carboxylic Acid, (4R)-2-Oxothiazolidine-4-carboxylic Acid, (R)-2-Oxothiazolidine-4-carboxylic Acid, 2-Oxo-L-thiazolidine-4-carboxylic Acid, L-2-Oxo-4-thiazolidinecarboxylic Acid, Procysteine, R-2-Oxo-1,3-thiazolidine-4-carboxylic Acid
InChI formel InChI=1S/C4H5NO3S/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)/t2-/m0/s1
LEDER OC(=O)[C@@H]1CSC(=O)N1
IUPAC-namn (4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid
Molekylvikt (g/mol) 147.15
Formel vikt 146.999