missing translation for 'onlineSavingsMsg'
Läs mer

(R)-2-Pyrrolidinone-5-carboxylic Acid, TRC

Produktkod. 30425912
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425912 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30425912 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9985001G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2-Pyrrolidinone-(R)-5-carboxylic Acid
CAS 4042-36-8
Molekylformel C5 H7 N O3
Kvantitet 1 g
Synonym D-Proline','5-oxo- (9CI','ACI), 5-Oxo-D-proline (ACI), Proline','5-oxo-','D- (8CI), (+)-2-Pyrrolidone-5-carboxylic acid, (+)-D-Pyroglutamic acid, (+)-Pyroglutamic acid, (2R)-2-Carboxy-5-pyrrolidinone, (2R)-5-Oxopyrrolidine-2-carboxylic acid, (R)-(+)-2-Pyrrolidone-5-carboxylic acid, (R)-(+)-2-Pyrrolidone-5-carboxylic acid, (R)-(+)-5-Oxopyrrolidine-2-carboxylic acid, (R)-2-Pyrrolidone-5-carboxylic acid, (R)-5-Oxoproline, (R)-5-Oxopyrrolidine-2-carboxylic acid, (R)-Pyroglutamic acid, D-2-Pyrrolidone-5-carboxylic acid, D-Pidolic acid, D-Pyroglutamic acid, D-Pyrrolidinonecarboxylic acid, D-Pyrrolidonecarboxylic acid
InChI formel InChI=1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m1/s1
LEDER OC(=O)[C@H]1CCC(=O)N1
IUPAC-namn (2R)-5-oxopyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 129.11
Formel vikt 129.0426
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.