Läs mer
(R)-2-((tert-Butoxycarbonyl)amino)-4-methylpentanoic Acid Hydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (R)-2-((tert-Butoxycarbonyl)amino)-4-methylpentanoic Acid Hydrate |
| CAS | 200937-17-3 |
| Molekylformel | C11H21NO4 . H2O |
| Kvantitet | 5 g |
| Synonym | D-Leucine, N-[(1,1-dimethylethoxy)carbonyl]-, hydrate (1:1) (ACI), D-Leucine, N-[(1,1-dimethylethoxy)carbonyl]-, monohydrate (9CI), (R)-2-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid hydrate, N-[(1,1-Dimethylethoxy)carbonyl]-D-leucine hydrate, (2R)-4-Methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid monohydrate, D-Boc leucine hydrate, N-(tert-Butoxycarbonyl)-D-leucine hydrate, N-Boc-D-leucine hydrate, tert-Butoxycarbonyl-D-leucine hydrate |
| LEDER | O.CC(C)C[C@@H](NC(=O)OC(C)(C)C)C(=O)O |
| Formel vikt | 249.1576 |