Läs mer
(R)-2-Thioxothiazolidine-4-carboxylic Acid-13C3, TRC
Produktkod. 30009364
Change view
Kvantitet:
0.5 mg
1 mg
10 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009364 1 mg -
30005441 0.5 mg 0.5mg
30008913 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3950021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2704345-00-4
Molekylformel C13C3H5NO2S2
Molekylvikt (g/mol) 166.2
Synonym (R)-2-Thioxo-4-thiazolidinecarboxylic Acid-13C3, (R)-Thiazolidine-2-thione-4-carboxylic Acid-13C3, (R)-TTCA-13C3, (R)-TTCA-13C3 (Thiram Metabolite), Raphanusamic Acid-13C3
IUPAC-namn (R)-2-thioxothiazolidine-4-carboxylic-4,5-13C2 acid-13C
LEDER S=C1N[13C@H]([13C](O)=O)[13CH2]S1
Specifikationer
Kemiskt namn eller material (R)-2-Thioxothiazolidine-4-carboxylic Acid-13C3
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C4H5NO2S2/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)/t2-/m0/s1/i1+1,2+1,3+1
Formel vikt 165.99
Procent renhet >95%