Läs mer
(R)-2-(Trifluoromethylsulfonyloxy)propionic Acid Methyl Ester, TRC
Produktkod. 30449040
Change view
Kvantitet:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449040 25 g
30452546 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79176025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-2-(Trifluoromethylsulfonyloxy)propionic Acid Methyl Ester
CAS 115724-80-6
Molekylformel C9 H14 O2
Kvantitet 25 g
Synonym (R)-2-(Trifluoromethylsulfonyloxy)-propionic Acid Methyl Ester, (2R)-2-(Trifluoromethylsulfonyloxy)-propionic Acid Methyl Ester
InChI formel InChI=1S/C9H14O2/c1-4-8(3)6-7-9(10)11-5-2/h4,6-7H,5H2,1-3H3/b7-6+,8-4+
LEDER CCOC(=O)\C=C\C(=C\C)\C
IUPAC-namn ethyl (2E,4E)-4-methylhexa-2,4-dienoate
Molekylvikt (g/mol) 154.206
Formel vikt 154.099