Läs mer
(R)-3-(4-Chlorophenyl)-4-hydroxybutyric Acid Sodium Salt, TRC
Produktkod. 30366161
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30366161 25 mg
1 options
Produktkod. 30366161 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37952825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-3-(4-Chlorophenyl)-4-hydroxybutyric Acid Sodium Salt
CAS 1346617-22-8
Molekylformel C10H10ClNaO3
Kvantitet 25 mg
Synonym (R)-4-Chloro-β-(hydroxymethyl)-benzenepropanoic Acid Sodium Salt, (R)-β-(p-Chlorophenyl)-γ-hydroxybutyric Acid Sodium Salt
InChI formel InChI=1S/C10H11ClO3.Na/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14;/h1-4,8,12H,5-6H2,(H,13,14);/q;+1/p-1/t8-;/m0./s1
LEDER ClC1=CC=C([C@@H](CC(O[Na])=O)CO)C=C1
IUPAC-namn sodium;(3R)-3-(4-chlorophenyl)-4-hydroxybutanoate
Molekylvikt (g/mol) 236.63