missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Thiazoles are used in peptide research. Mainly used in medicine field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiazoles are used in peptide research. Mainly used in medicine field.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
Thiazoles are used in peptide research. Mainly used in medicine field.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
Specifications
Melting Point | 100°C to 107°C |
Color | White to Brown |
Quantity | 250 mg |
Formula Weight | 237.27 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | (R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'