missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Bromo-alpha-methylbenzyl alcohol, 95%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-3211067
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
134615-24-0
C8H9BrO
201.06
MFCD06658929
ULMJQMDYAOJNCC-ZCFIWIBFSA-N
r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethan-1-ol, pubchem15565, r-1-m-bromophenyl ethanol, r-1-3-bromo-phenyl-ethanol, r-3-bromo-alpha-methylbenzyl alcohol, benzenemethanol, 3-bromo-a-methyl-, ar
2734889
(1R)-1-(3-bromophenyl)ethanol
CC(C1=CC(=CC=C1)Br)O
This item is not returnable. View return policy
134615-24-0
C8H9BrO
201.06
MFCD06658929
ULMJQMDYAOJNCC-ZCFIWIBFSA-N
r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethan-1-ol, pubchem15565, r-1-m-bromophenyl ethanol, r-1-3-bromo-phenyl-ethanol, r-3-bromo-alpha-methylbenzyl alcohol, benzenemethanol, 3-bromo-a-methyl-, ar
2734889
(1R)-1-(3-bromophenyl)ethanol
CC(C1=CC(=CC=C1)Br)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)C6H4Br
Quantity 1 g
Formula Weight 201.06
Percent Purity 95%,98% ee
Physical Form Liquid
Chemical Name or Material (R)-3-Bromo-α-methylbenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-3-Bromo-a-methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'