Läs mer
(R)-3-Hydroxy-2-phenylpropanoic acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (R)-3-Hydroxy-2-phenylpropanoic acid |
| CAS | 17126-67-9 |
| Molekylformel | C9H10O3 |
| Kvantitet | 50 mg |
| Synonym | (αR)-α-(Hydroxymethyl)-benzeneacetic acid, Benzeneacetic acid, α-(hydroxymethyl)-, (R)-, Tropic acid, (R)-(+)- (8CI), (+)-(R)-Tropic acid, (+)-3-Hydroxy-2-phenylpropionic acid, (+)-Tropic acid, (+)-α-(Hydroxymethyl)benzeneacetic acid, (R)-(+)-Tropic acid, (R)-Tropic acid, D-(+)-Tropic acid, D-Tropic acid |
| InChI formel | InChI=1S/C9H10O3/c10-6-8(9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)/t8-/m0/s1 |
| LEDER | OC[C@H](C(=O)O)c1ccccc1 |
| IUPAC-namn | (2R)-3-hydroxy-2-phenylpropanoic acid |
| Molekylvikt (g/mol) | 166.17 |
| Formel vikt | 166.063 |