Läs mer
(R)-(+)-3-Methylcyclopentanone, TRC
Produktkod. 30472943
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472943 500 mg
30469718 250 mg
30470372 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472943 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM479038500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6672-30-6
Molekylformel C6H10O
Molekylvikt (g/mol) 98.14
Synonym (3R)-3-Methylcyclopentanone, (+)-3-Methylcyclopentanone, (3R)-3-Methylcyclopentan-1-one, (R)-3-Methylcyclopentanone
IUPAC-namn (3R)-3-methylcyclopentan-1-one
LEDER C[C@@H]1CCC(=O)C1
Specifikationer
Kemiskt namn eller material (R)-(+)-3-Methylcyclopentanone
Kvantitet 500 mg
InChI formel InChI=1S/C6H10O/c1-5-2-3-6(7)4-5/h5H,2-4H2,1H3/t5-/m1/s1
Formel vikt 98.0732