Läs mer
(R)-4-Amino-3-hydroxybutanoic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (R)-4-Amino-3-hydroxybutanoic Acid |
| CAS | 7013-07-2 |
| Molekylformel | C4H9NO3 |
| Kvantitet | 100 mg |
| Synonym | (3R)-4-Amino-3-hydroxybutanoic Acid, (-)-β-Hydroxy-GABA, (-)-γ-Amino-β-hydroxybutyric Acid, (3R)-(-)-4-Amino-3-hydroxybutanoic Acid, (3R)-4-Amino-3-hydroxybutanoic Acid, (3R)-4-Azaniumyl-3-hydroxybutanoate, (R)-(-)-3-Hydroxy-GABA, (R)-(-)-β-Hydroxy-GABA, (R)-4-Amino-3-hydroxybutanoic Acid, (R)-4-Amino-3-hydroxybutanoic Acid, (R)-4-Amino-3-hydroxybutyric Acid, (R)-GABOB, L-γ-Amino-β-hydroxybutyric Acid, R-(-)-γ-Amino-β-hydroxybutyric Acid |
| InChI formel | InChI=1S/C4H9NO3/c5-2-3(6)1-4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m1/s1 |
| LEDER | NC[C@H](O)CC(=O)O |
| IUPAC-namn | (3R)-4-amino-3-hydroxybutanoic acid |
| Molekylvikt (g/mol) | 119.12 |
| Formel vikt | 119.0582 |