Learn More
Alfa Aesar™ (R)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%
Brand:  Alfa Aesar™ H63322.03
Additional Details : CAS Number : 387824-79-5 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15488236
  • 2632.13 SEK / 1g
Estimated Shipment date
from Supplier 07-03-2018
Add to basket
Specifications
Chemical Identifiers
(R)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid
474.51
Fmoc-D-Dab(Z)-OH
387824-79-5
MFCD02684372
Specifications
(R)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid
Powder
474.51
White
Fmoc-D-Dab(Z)-OH
387824-79-5
95%
MFCD02684372
1g