Läs mer
(R)-4-Bromo Phenylephrine Hydrochloride, TRC
Produktkod. 30330886
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30330886 1 mg
1 options
Produktkod. 30330886 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6868351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-4-Bromo Phenylephrine Hydrochloride
CAS 1391068-15-7
Molekylformel C9H13BrClNO2
Kvantitet 1 mg
Synonym (R)-4-Bromo-3-hydroxy-alpha-[(methylamino)methyl]benzenemethanol Hydrochloride, (R)4-Bromophenylephrine Hydrochloride
InChI formel InChI=1S/C9H12BrNO2.ClH/c1-11-5-9(13)6-2-3-7(10)8(12)4-6;/h2-4,9,11-13H,5H2,1H3;1H/t9-;/m0./s1
LEDER Cl.CNC[C@H](O)c1ccc(Br)c(O)c1
IUPAC-namn 2-bromo-5-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
Molekylvikt (g/mol) 282.56
Formel vikt 280.9818