Läs mer
(R)-6-Bromo Phenylephrine Hydrochloride, TRC
Produktkod. 30346955
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30346955 10 mg
1 options
Produktkod. 30346955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68683010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-6-Bromo Phenylephrine Hydrochloride
CAS 1391067-95-0
Molekylformel C9H13BrClNO2
Kvantitet 10 mg
Synonym (R)-6-Bromo-3-hydroxy-alpha-[(methylamino)methyl]benzenemethanol Hydrochloride, (R)-6-Bromophenylephrine Hydrochloride
InChI formel InChI=1S/C9H12BrNO2.ClH/c1-11-5-9(13)7-4-6(12)2-3-8(7)10;/h2-4,9,11-13H,5H2,1H3;1H/t9-;/m0./s1
LEDER Cl.CNC[C@H](O)c1cc(O)ccc1Br
IUPAC-namn 4-bromo-3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
Molekylvikt (g/mol) 282.56
Formel vikt 280.9818