Läs mer
(R)-Albuterol Hydrochloride, TRC
Produktkod. 30134382
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134382 50 mg
17954745 5 mg
17902785 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA51448550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50293-90-8
Molekylformel C13 H21 N O3 . Cl H
Molekylvikt (g/mol) 275.77
Synonym Levosalbutamol hydrochloride, Levalbuterol hydrochloride, (R)-Albuterol hydrochloride, 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, hydrochloride (1:1), (α1R)-, 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, hydrochloride, (R)-, 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, hydrochloride, (α1R)- (9CI), (-)-Salbutamol hydrochloride, (R)-4-(2-(tert-Butylamino)-1-hydroxyethyl)-2-(hydroxymethyl)phenol hydrochloride, (R)-Salbutamol hydrochloride, Xopenex
IUPAC-namn 4-[(1R)-2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;hydrochloride
LEDER Cl.CC(C)(C)NC[C@H](O)c1ccc(O)c(CO)c1
Specifikationer
Kemiskt namn eller material Levalbuterol hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C13H21NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,12,14-17H,7-8H2,1-3H3;1H/t12-;/m0./s1
Formel vikt 275.1288