Läs mer
(R)-alpha-[(2,2-Dimethyl-1-oxopropyl)amino]-4-hydroxybenzeneacetic Acid, TRC
Produktkod. 30136367
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136367 10 g
30008526 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136367 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47655510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 205826-86-4
Molekylformel C13 H17 N O4
Molekylvikt (g/mol) 251.28
Synonym (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid, Amoxicillin Trihydrate Imp. H (EP)
IUPAC-namn (2R)-2-(2,2-dimethylpropanoylamino)-2-(4-hydroxyphenyl)acetic acid
LEDER CC(C)(C)C(=O)N[C@@H](C(=O)O)c1ccc(O)cc1
Specifikationer
Kemiskt namn eller material (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C13H17NO4/c1-13(2,3)12(18)14-10(11(16)17)8-4-6-9(15)7-5-8/h4-7,10,15H,1-3H3,(H,14,18)(H,16,17)/t10-/m1/s1
Formel vikt 251.1158
Procent renhet >95%