Läs mer
(R)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid, TRC
Produktkod. 30376994
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376994 25 g
30371678 1 g
30371796 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30376994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE91697025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63422-71-9
Molekylformel C15 H17 N3 O5
Molekylvikt (g/mol) 319.31
Synonym (2R)-2-[[(4-Ethyl-2,3-dioxopiperazin-1-yl)carbonyl]amino]-2-phenylacetic Acid, Piperacillin Sodium Imp. G (EP)
IUPAC-namn (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetic acid
LEDER CCN1CCN(C(=O)N[C@@H](C(=O)O)c2ccccc2)C(=O)C1=O
Specifikationer
Kemiskt namn eller material (2R)-2-[((4-Ethyl-2,3-dioxopiperazin-1-yl)carbonyl)amino]-2-phenylacetic Acid
Kvantitet 25 g
InChI formel InChI=1S/C15H17N3O5/c1-2-17-8-9-18(13(20)12(17)19)15(23)16-11(14(21)22)10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3,(H,16,23)(H,21,22)/t11-/m1/s1
Formel vikt 319.1168