Läs mer
R(-)-Alpha-Chloromethyl Histamine Dihydrochloride, TRC
Produktkod. 30363423
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30363423 250 mg
1 options
Produktkod. 30363423 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC369535250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material R(-)-alpha-Chloromethyl Histamine Dihydrochloride
CAS 75614-86-7
Molekylformel C6H12Cl3N3
Kvantitet 250 mg
Synonym α-(Chloromethyl)-1H-imidazole-4-ethanamine Dihydrochloride
InChI formel InChI=1S/C6H10ClN3.2ClH/c7-2-5(8)1-6-3-9-4-10-6;;/h3-5H,1-2,8H2,(H,9,10);2*1H/t5-;;/m0../s1
LEDER N[C@H](CCl)CC1=CNC=N1
IUPAC-namn (S)-1-chloro-3-(1H-imidazol-4-yl)propan-2-amine di hydrogen chloride
Molekylvikt (g/mol) 232.54
Formel vikt 231.01