Läs mer
(R)-(+)-alpha-Lipoic Acid, TRC
Produktkod. 30135800
Change view
Kvantitet:
50 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135800 500 mg
30132584 50 mg
30012112 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135800 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL468725500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1200-22-2
Molekylformel C8 H14 O2 S2
Molekylvikt (g/mol) 206.33
Synonym Thioctic Acid
IUPAC-namn 5-[(3R)-dithiolan-3-yl]pentanoic acid
LEDER OC(=O)CCCC[C@@H]1CCSS1
Specifikationer
Kemiskt namn eller material (+)-a-Lipoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)/t7-/m1/s1
Formel vikt 206.0435
Procent renhet >95%