missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride monohydrate, 97%, Thermo Scientific™

Catalog Number p-4229054
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104347-13-9
C4H7NO2·ClH
155.59
XBKCXPRYTLOQKS-AENDTGMFSA-N
d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 3r-3-aminotetrahydrofuran-2-one hydrochloride, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-a-amino-g-butyrolactone hydrochloride, pubchem14743, r-+-, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl
11954435
(3R)-3-aminooxolan-2-one;hydrochloride
C1COC(=O)C1N.Cl
This item is not returnable. View return policy
104347-13-9
C4H7NO2·ClH
155.59
XBKCXPRYTLOQKS-AENDTGMFSA-N
d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 3r-3-aminotetrahydrofuran-2-one hydrochloride, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-a-amino-g-butyrolactone hydrochloride, pubchem14743, r-+-, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl
11954435
(3R)-3-aminooxolan-2-one;hydrochloride
C1COC(=O)C1N.Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 215.1°C to 217.1°C
Color Yellow to Brown
Elemental Analysis 29.64 to 32.12% C, 8.64 to 9.36% N
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Specific Rotation 30
Formula Weight 155.59
Percent Purity 97%
Physical Form Crystalline Powder, Powder, or Solid
Chemical Name or Material (R)-(+)-α-Amino-γ-butyrolactone hydrochloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'