Läs mer
(R)-Amygdalin, TRC
Produktkod. 30313425
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30313425 25 g
1 options
Produktkod. 30313425 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA57684025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amygdalin
CAS 29883-15-6
Molekylformel C20 H27 N O11
Kvantitet 25 g
Synonym Benzeneacetonitrile, α-[(6-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-, (αR)-, Amygdalin (6CI,7CI,8CI), Benzeneacetonitrile, α-[(6-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-, (R)-, (αR)-α-[(6-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]benzeneacetonitrile, (-)-Amygdalin, (R)-Amygdalin, Amygdaloside, Amygdaloside (disaccharide), D-Amygdalin, Mandelonitrile-β-gentiobioside, NSC 15780
InChI formel InChI=1S/C20H27NO11/c21-6-10(9-4-2-1-3-5-9)30-20-18(28)16(26)14(24)12(32-20)8-29-19-17(27)15(25)13(23)11(7-22)31-19/h1-5,10-20,22-28H,7-8H2/t10-,11+,12+,13+,14+,15-,16-,17+,18+,19+,20+/m0/s1
LEDER OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](O[C@@H](C#N)c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2R)-2-phenyl-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyacetonitrile
Molekylvikt (g/mol) 457.43
Formel vikt 457.1584