missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-beta-metylfenetylamin, TRC

Code produit 30332781
Modifier l'affichage
Click to view available options
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Kvantitet
30332781 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30332781 Fournisseur Toronto Research Chemicals Code fournisseur TRCB3999385G
 Begränsad artikel

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (R)-(+)-beta-methylphenethylamine
CAS 28163-64-6
Molekylformel C9 H13 N
Kvantitet 5 g
Synonym (βR)-β-Methylbenzeneethanamine, ((R)-2-Phenylpropan-1-yl)amine, (+)-2-Phenylpropylamine, (+)-β-Methylphenethylamine, (2R)-1-Amino-2-phenylpropane, (2R)-2-Phenylpropan-1-amine, (2R)-2-Phenylpropan-1-amine, (2R)-2-Phenylpropylamine, (R)-(+)--Methylphenethylamine, (R)-(+)-2-Phenylpropylamine, (R)-2-Phenyl-1-propanamine, (R)-2-Phenyl-1-propylamine, (R)-2-Phenylpropylamine, (R)-2-Phenylpropylamine, (R)-β-Methylphenethylamine
InChI formel InChI=1S/C9H13N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/t8-/m0/s1
LEDER C[C@@H](CN)c1ccccc1
IUPAC-namn (2R)-2-fenylpropan-1-amin
Molekylvikt (g/mol) 135.21
Formel vikt 135.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.