Läs mer
(R)-Bethanechol, TRC
Produktkod. 30332807
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30332807 10 mg
30333289 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30332807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB32889010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Bethanechol
CAS 944538-49-2
Molekylformel C7H17ClN2O2
Kvantitet 10 mg
Synonym (2R)-2-[(Aminocarbonyl)oxy]-N,N,N-trimethyl-1propanaminium Chloride
InChI formel InChI=1S/C7H16N2O2.ClH/c1-6(11-7(8)10)5-9(2,3)4;/h6H,5H2,1-4H3,(H-,8,10);1H/t6-;/m1./s1
LEDER [Cl-].C[C@H](C[N+](C)(C)C)OC(=O)N
IUPAC-namn [(2R)-2-carbamoyloxypropyl]-trimethylazanium;chloride
Molekylvikt (g/mol) 196.68
Formel vikt 196.0979