Läs mer
(R)-Bicalutamide, TRC
Produktkod. 30345091
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30345091 25 mg
1 options
Produktkod. 30345091 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB38200525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Bicalutamide
CAS 113299-40-4
Molekylformel C18 H14 F4 N2 O4 S
Kvantitet 25 mg
Synonym Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methyl-, (2R)-, Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methyl-, (R)-, (2R)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide, (R)-Bicalutamide, (R)-Casodex
InChI formel InChI=1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)/t17-/m0/s1
LEDER C[C@](O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
IUPAC-namn (2R)-N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
Molekylvikt (g/mol) 430.37
Formel vikt 430.061