Läs mer
(R)-Bis(1-methylethyl) Ester [[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid, TRC
Produktkod. 30314430
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30314430 250 mg
1 options
Produktkod. 30314430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA123895250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diisopropyl ([(2R)-1-(6-Amino-9H-purin-9-yl)propan-2-yloxy]methyl)phosphonate
CAS 160616-04-6
Molekylformel C15 H26 N5 O4 P
Kvantitet 250 mg
Synonym P-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid Bis(1-methylethyl) Ester, Phosphonic acid, [[(1R)-2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, bis(1-methylethyl) ester (9CI), Phosphonic acid, [[2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, bis(1-methylethyl) ester, (R)-, 9-[(2R)-2-[[(Diisopropyloxy)phosphoryl]methoxy]propyl]-9H-purin-6-amine, Diisopropyl [[(2R)-1-(6-Amino-9H-purin-9-yl)propan-2-yloxy]methyl]phosphonate
InChI formel InChI=1S/C15H26N5O4P/c1-10(2)23-25(21,24-11(3)4)9-22-12(5)6-20-8-19-13-14(16)17-7-18-15(13)20/h7-8,10-12H,6,9H2,1-5H3,(H2,16,17,18)/t12-/m1/s1
LEDER CC(C)OP(=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)OC(C)C
IUPAC-namn 9-[(2R)-2-[di(propan-2-yloxy)phosphorylmethoxy]propyl]purin-6-amine
Molekylvikt (g/mol) 371.3718
Formel vikt 371.1722