Läs mer
(R)-(+)-Blebbistatin, Tocris Bioscience™
Beskrivning
Inactive enantiomer of the selective inhibitor of myosin II (±)-blebbistatin.
Active Enantiomer and Racemate also available.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (R)-(+)-Blebbistatin |
| CAS | 1177356-70-5 |
| Molekylformel | C18H16N2O2 |
| Kvantitet | 1 mg |
| Synonym | +-blebbistatin, r-+-blebbistatin, blebbistatin, +, unii-xwg1958e8i, insolutionblebbistatin, racemic, r-blebbistatin, +-brebbistatin, tocris-1760, 4h-pyrrolo 2,3-b quinolin-4-one, 1,2,3,3a-tetrahydro-3a-hydroxy-6-methyl-1-phenyl-, 3ar, unii-20wc4j7cq6 component |
| InChI-nyckel | LZAXPYOBKSJSEX-SFHVURJKSA-N |
| LEDER | CC1=CC2=C(C=C1)N=C3C(C2=O)(CCN3C4=CC=CC=C4)O |
| IUPAC-namn | (3aR)-3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-one |
| Molekylvikt (g/mol) | 292.338 |
| PubChem CID | 6604910 |
| Visa mer |