Läs mer
(R)-(+)-Bupivacaine Hydrochloride, TRC
Produktkod. 30137517
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137517 25 mg
30007002 250 mg
30011878 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137517 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68954525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27262-46-0
Molekylformel C18 H28 N2 O . HCl
Molekylvikt (g/mol) 324.89
Synonym 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, hydrochloride (1:1), (2R)-, 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, monohydrochloride, (2R)- (9CI), 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, monohydrochloride, (R)-, 2',6'-Pipecoloxylidide, 1-butyl-, monohydrochloride, (+)- (8CI), (+)-Bupivacaine monohydrochloride, (R)-(+)-Bupivacaine monohydrochloride, (R)-Bupivacaine hydrochloride, (2R)-1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide hydrochloride, (R)-Bupivacaine HCl
IUPAC-namn (2R)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride
LEDER Cl.CCCCN1CCCC[C@@H]1C(=O)Nc2c(C)cccc2C
Specifikationer
Kemiskt namn eller material (R)-(+)-Bupivacaine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C18H28N2O.ClH/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H/t16-;/m1./s1
Formel vikt 324.1968
Procent renhet >95%