Läs mer
(R)-(+)-Canadine, TRC
Produktkod. 30330750
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30330750 100 mg
1 options
Produktkod. 30330750 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC175170100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-(+)-Canadine
CAS 2086-96-6
Molekylformel C20H21NO4
Kvantitet 100 mg
Synonym (R)-5,8,13,13a-Tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, (+)-Canadine, (+)-Tetrahydroberberine, (R)-(+)-7,8,13,13a-Tetrahydroberberine, 9,10-Dimethoxy-2,3-(methylenedioxy)-13abeta-berbine, d-Canadine, beta-Canadine
InChI formel InChI=1S/C20H21NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,8-9,16H,5-7,10-11H2,1-2H3/t16-/m1/s1
LEDER COc1ccc2C[C@H]3N(CCc4cc5OCOc5cc34)Cc2c1OC
IUPAC-namn (1R)-16,17-dimethoxy-5,7-dioxa-13-azapentacyclo[11.8.0.02,10.04,8.015,20]henicosa-2,4(8),9,15(20),16,18-hexaene
Molekylvikt (g/mol) 339.39
Formel vikt 339.1471