Läs mer
R-(+)-Carbidopa, TRC
Produktkod. 30365781
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365781 5 mg
30363099 10 mg
30353095 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365781 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1759865MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28875-92-5
Molekylformel C10 H14 N2 O4
Molekylvikt (g/mol) 226.23
Synonym Benzenepropanoic acid, α-hydrazinyl-3,4-dihydroxy-α-methyl-, (αR)-, Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, (R)-, Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, (αR)- (9CI), Hydrocinnamic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, D- (8CI), (αR)-α-Hydrazinyl-3,4-dihydroxy-α-methylbenzenepropanoic acid, MK 485, (2R)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic Acid, (R)-Carbidopa, ent-Carbidopa
IUPAC-namn (2R)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid
LEDER C[C@](Cc1ccc(O)c(O)c1)(NN)C(=O)O
Specifikationer
Kemiskt namn eller material R-(+)-Carbidopa
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C10H14N2O4/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6/h2-4,12-14H,5,11H2,1H3,(H,15,16)/t10-/m1/s1
Formel vikt 226.0954
Procent renhet >95%