Läs mer
(R)-Carisbamate, TRC
Produktkod. 30353090
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353090 5 mg
30364915 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353090 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1835055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 194085-74-0
Molekylformel C9 H10 Cl N O3
Molekylvikt (g/mol) 215.63
Synonym 1,2-Ethanediol, 1-(2-chlorophenyl)-, 2-carbamate, (1R)-, 1,2-Ethanediol, 1-(2-chlorophenyl)-, 2-carbamate, (R)-, (R)-Carisbamate
IUPAC-namn [(2R)-2-(2-chlorophenyl)-2-hydroxyethyl] carbamate
LEDER NC(=O)OC[C@H](O)c1ccccc1Cl
Specifikationer
Kemiskt namn eller material (R)-Carisbamate
Kvantitet 5 mg
InChI formel InChI=1S/C9H10ClNO3/c10-7-4-2-1-3-6(7)8(12)5-14-9(11)13/h1-4,8,12H,5H2,(H2,11,13)/t8-/m0/s1
Formel vikt 215.0349