Läs mer
(R)-(+)-Celiprolol Hydrochloride, TRC
Produktkod. 30351109
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351109 1 mg
30357740 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30351109 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2544901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57470-78-7
Molekylformel C20 H33 N3 O4 . Cl H
Molekylvikt (g/mol) 415.95
Synonym Celiprolol Hydrochloride, Celectol, Corliprol, NSC 324509, REV 5320A, ST 1396, Selecor, Urea, N'-[3-acetyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethyl-, monohydrochloride (9CI)
IUPAC-namn 3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride
LEDER Cl.CCN(CC)C(=O)Nc1ccc(OCC(O)CNC(C)(C)C)c(c1)C(=O)C
Specifikationer
Kemiskt namn eller material Celiprolol Hydrochloride
Kvantitet 1 mg
InChI formel InChI=1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H
Formel vikt 415.2238