missing translation for 'onlineSavingsMsg'
Läs mer

(R)-Chloropheniramine 3,5-Dinitrobenzoic Acid, TRC

Produktkod. 30351098
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351098 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30351098 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC42429825MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (R)-Chloropheniramine 3,5-Dinitrobenzoic Acid
CAS 1180493-26-8
Molekylformel C16H19ClN2 . C7H4N2O6
Kvantitet 25 mg
Synonym (gammaR)-gamma-(4-Chlorophenyl)-N,N-dimethyl-2-pyridinepropanamine 3,5-Dinitrobenzoic Acid
InChI formel InChI=1S/C16H19ClN2.C7H4N2O6/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h3-9,11,15H,10,12H2,1-2H3;1-3H,(H,10,11)/t15-;/m1./s1
LEDER CN(C)CC[C@H](c1ccc(Cl)cc1)c2ccccn2.OC(=O)c3cc(cc(c3)[N+](=O)[O-])[N+](=O)[O-]
IUPAC-namn (3R)-3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine;3,5-dinitrobenzoic acid
Molekylvikt (g/mol) 486.9
Formel vikt 486.1306
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.