Läs mer
(R)-(-)-Chlorpheniramine Maleate, TRC
Produktkod. 30009199
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009199 1 mg
30004038 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009199 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4243061MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23095-76-3
Molekylformel C16H19ClN2 • C4H4O4
Molekylvikt (g/mol) 274.7911607
Synonym (γR)-γ-(4-chlorophenyl)-N,N-dimethyl-2-Pyridinepropanamine (2Z)-2-butenedioate (1:1), (R)-γ-(4-chlorophenyl)-N,N-dimethyl-2-Pyridinepropanamine (Z)-2-butenedioate (1:1), (-)-2-[p-chloro-α-[2-(dimethylamino)ethyl]benzyl]-Pyridine maleate (1:1), (-)-Chlorpheniramine maleate, (R)-γ-(4-chlorophenyl)-N,N-dimethyl-2-Pyridinepropanamine (2Z)-2-butenedioate (1:1), L-Chlorpheniramine maleate
IUPAC-namn (Z)-but-2-enedioic acid;(3R)-3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine
LEDER CN(C)CC[C@H](C1=CC=C(C=C1)Cl)C2=CC=CC=N2.C(=C\C(=O)O)\C(=O)O
Specifikationer
Kemiskt namn eller material (R)-(-)-Chlorpheniramine Maleate
Kvantitet 1 mg
InChI formel InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-;/m1./s1