Läs mer
Tocris Bioscience™ (R)-Citalopram oxalate
GREENER_CHOICE
Produktkod. 18743989
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18743989 50 mg
18707978 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18743989 Leverantör Tocris Bioscience™ Leverantörsnummer 5763/50
Enantiomer of escitalopram oxalate (Cat. No. 1427)
(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.
Racemate and S-enantiomer also available.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate
CAS 219861-53-7
Kvantitet 50 mg
Mål 5-HT Transporter Inhibitors
Molekylformel C20H21FN2O.C2H2O4
Formel vikt 414.43
Renhet >98%