Läs mer
(R)-Citalopram Oxalate, TRC
Produktkod. 30138292
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138292 100 mg
30011416 25 mg
17993065 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC505015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 219861-53-7
Molekylformel C20 H21 F N2 O . C2 H2 O4
Molekylvikt (g/mol) 414.43
Synonym (R)-Citalopram oxalate, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, (1R)-, ethanedioate (1:1), (R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile ethanedioate, R-(-)-Citalopram oxalate, Orlistat Related Compound A (USP)
IUPAC-namn (1R)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
LEDER CN(C)CCC[C@@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O
Specifikationer
Kemiskt namn eller material (R)-Citalopram Oxalate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m1./s1
Formel vikt 414.1591
Procent renhet >95%