Läs mer
Tocris Bioscience™ (R)-Citalopram oxalate
Enantiomer of escitalopram oxalate (Cat. No. 1427)
Brand: Tocris Bioscience™ 5763/10
Beskrivning
(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.Racemate and S-enantiomer also available.
Specifikationer
| (-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate | |
| 219861-53-7 | |
| 10 mg |