Läs mer
(R)-Coniine Hydrochloride (~25% S-isomer), TRC
Produktkod. 30137319
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137319 25 mg
30132714 250 mg
30006309 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137319 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70104025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 88057-03-8
Molekylformel C8 H17 N . H Cl
Molekylvikt (g/mol) 163.69
Synonym (R)-2-Propylpiperidine Hydrochloride, (-)-Coniine hydrochloride
IUPAC-namn (2R)-2-propylpiperidine;hydrochloride
LEDER Cl.CCC[C@@H]1CCCCN1
Specifikationer
Kemiskt namn eller material (R)-Coniine hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C8H17N.ClH/c1-2-5-8-6-3-4-7-9-8;/h8-9H,2-7H2,1H3;1H/t8-;/m1./s1
Formel vikt 163.1128