Läs mer
(R)-D-Cyclohexylalanine, TRC
Produktkod. 30363713
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30363713 5 g
1 options
Produktkod. 30363713 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9860705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-D-Cyclohexylalanine
CAS 58717-02-5
Molekylformel C9 H17 N O2
Kvantitet 5 g
Synonym C9H17NO2
InChI formel InChI=1S/C9H17NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h7-8H,1-6,10H2,(H,11,12)/t8-/m1/s1
LEDER N[C@H](CC1CCCCC1)C(=O)O
IUPAC-namn (2R)-2-amino-3-cyclohexylpropanoic acid
Molekylvikt (g/mol) 171.237
Formel vikt 171.126