Läs mer
R&D Systems™ Caspase-1/ICE Inhibitor Z-WEHD-FMK
Kvantitet:
1 mg
Beskrivning
- Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying caspase activity
- Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
- Regulated by a variety of stimili, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin
Specifikationer
Specifikationer
| Innehåll och lagring | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Kvantitet | 1 mg |
| Produkttyp | Caspase-1 |
| Molekylvikt (g/mol) | 877Da |
| Inhibitorer | Z-WEHD-FMK |
| Form | Lyophilized |
For research use only. Not for use in diagnostic procedures