Läs mer
Tocris Bioscience™ CU-T12-9
Beskrivning
CU-T12-9 is a potent TLR1/2 agonist (EC50 = 52.9 nM). Activates the NF-κB pathway and upregulates proinflammatory cytokines in vitro. Also enhances TLR1 and TLR2 dimerization.
Specifikationer
Specifikationer
| Kemiskt namn eller material | CU-T12-9 |
| CAS | 1821387-73-8 |
| Synonym | N-Methyl-4-nitro-2-(4-(4-trifluoromethyl)phenyl)-1H-imidazol-1-yl)aniline |
| Kvantitet | 10 mg |
| Löslighet | Soluble to 100mM in DMSO and to 20mM in ethanol |
| Molekylformel | C17H13F3N4O2 |
| Formel vikt | 362.31 |
| Fysisk form | Solid |
| Produkttyp | Bioactive Small Molecule |
| Renhet | >99% |
Research Use Only. Not For Human or Veterinary Use