Läs mer
Tocris Bioscience™ Deltarasin
High affinity PDEδ-KRAS interaction inhibitor
Brand: Tocris Bioscience™ 5424/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Deltarasin is a high affinity PDEδ-KRAS interaction inhibitor (Kd = 41 nM); binds to PDEδ. Reduces proliferation and induces cell death of KRAS-dependent Panc-Tu-1 and Capan-1 pancreatic cancer cells. Attenuates tumor growth of Panc-Tu-1 xenografts in mice.Specifikationer
| Other Ras GTPase Compounds | |
| 2-[4-[(2S)-2-(2-Phenyl-1H-benzimidazol-1-yl)-2-(4-piperidinyl)ethoxy]phenyl]-1-(phenylmethyl)-1H-benzimidazole trihydrochloride | |
| Soluble to 100mM in water and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 1440898-82-7 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use